Methyl acetoacetate, 99+%, Thermo Scientific Chemicals
Methyl acetoacetate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl acetoacetate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number 126150025
Price (USD)/ Each
85.65
Online exclusive
94.60 
Save 8.95 (9%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
85.65
Online exclusive
94.60 
Save 8.95 (9%)
Add to cart
Methyl acetoacetate, 99+%, Thermo Scientific Chemicals
Catalog number126150025
Price (USD)/ Each
85.65
Online exclusive
94.60 
Save 8.95 (9%)
-
Add to cart
Chemical Identifiers
IUPAC Namemethyl 3-oxobutanoate
Molecular FormulaC5H8O3
InChI KeyWRQNANDWMGAFTP-UHFFFAOYSA-N
SMILESCOC(=O)CC(C)=O
Molecular Weight (g/mol)116.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4175 to 1.4200 (20°C, 589 nm)
Free acid (CH3COOH)=<0.2 %
Color scale=<20 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only