1x1 image pixel for data collection
Methyl acrylate, 99%, stabilized
Methyl acrylate, 99%, stabilized
Methyl acrylate, 99%, stabilized
Methyl acrylate, 99%, stabilized
Thermo Scientific Chemicals

Methyl acrylate, 99%, stabilized

CAS: 96-33-3 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass bottle
Catalog number 126195000
Price (USD)
38.65
Special offer
Online exclusive
44.90
Save 6.25 (14%)
Each
In stock
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.65
Special offer
Online exclusive
44.90
Save 6.25 (14%)
Each
Add to cart
Chemical Identifiers
CAS96-33-3
IUPAC Namemethyl prop-2-enoate
Molecular FormulaC4H6O2
InChI KeyBAPJBEWLBFYGME-UHFFFAOYSA-N
SMILESCOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4010 to 1.4040 (20°C, 589 nm)
Color scale=<10 APHA
Infrared spectrumConforms
Water=<0.08 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection