N-Methyl-beta-alaninenitrile, 98%
N-Methyl-beta-alaninenitrile, 98%
Thermo Scientific Chemicals

N-Methyl-beta-alaninenitrile, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Packaging:
Glass bottle
Catalog number 126220050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-(methylamino)propanenitrile
Molecular FormulaC4H8N2
InChI KeyUNIJBMUBHBAUET-UHFFFAOYSA-N
SMILESCNCCC#N
Molecular Weight (g/mol)84.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Refractive index1.4300 to 1.4320 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only