N-Methylaniline, 99%
N-Methylaniline, 99%
N-Methylaniline, 99%
N-Methylaniline, 99%
Thermo Scientific Chemicals

N-Methylaniline, 99%

CAS: 100-61-8 | C7H9N | 107.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 126275000
Price (USD)
44.65
Online Exclusive
49.10
Save 4.45 (9%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.10
Save 4.45 (9%)
Each
Add to cart
Chemical Identifiers
CAS100-61-8
IUPAC NameN-methylaniline
Molecular FormulaC7H9N
InChI KeyAFBPFSWMIHJQDM-UHFFFAOYSA-N
SMILESCNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5685 to 1.5725 (20°C, 589 nm)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection