Thermo Scientific Chemicals

Methyl benzoate, 99%, Thermo Scientific Chemicals

Catalog number: 126340025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Methyl benzoate, 99%, Thermo Scientific Chemicals

Catalog number: 126340025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 126340025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-58-3
IUPAC Name
methyl benzoate
Molecular Formula
C8H8O2
InChI Key
QPJVMBTYPHYUOC-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1
Appearance (Color)
Clear colorless to light yellow
GC
>=98.5 %
Specific gravity
(25/25°C) 1.084 to 1.088
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text