Recovery™ Cell Culture Freezing Medium
Recovery™ Cell Culture Freezing Medium
Gibco™

Recovery™ Cell Culture Freezing Medium

Recovery Cell Culture Freezing Medium is a complete, ready-to-use freezing medium for cryopreservation of a wide variety of mammalian cells,Read more
Have Questions?
Catalog number 12648010
Price (USD)/ 50 mL
177.00
-
Add to cart
Price (USD)/ 50 mL
177.00
Add to cart
Recovery™ Cell Culture Freezing Medium
Catalog number12648010
Price (USD)/ 50 mL
177.00
-
Add to cart
Recovery Cell Culture Freezing Medium is a complete, ready-to-use freezing medium for cryopreservation of a wide variety of mammalian cells, including CHO-S, CHO-K1, HEK 293, Jurkat, and NIH 3T3. Recovery Cell Culture Freezing Medium features:

• Classic formulation optimized for better performance
• Suitability for a wide variety of mammalian cells
• Quality and performance tested

Classic formulation optimized for better performance
For many years, researchers have used a common recipe for making freezing medium: High-glucose Dulbecco's Modified Eagle Medium (DMEM) supplemented with 10% serum and 10% DMSO. Recovery Cell Culture Freezing Medium is an optimized version of this classic formulation, using high-quality Gibco Bovine Serum and Fetal Bovine Serum to provide improved cell recovery and viability after thawing.

Suitability for a wide variety of mammalian cells
Recovery Cell Culture Freezing Medium can be used to freeze most mammalian cells cultured in DMEM, DMEM/F-12, MEM, RPMI 1640, and Ham's F-12, including CHO-S, CHO-K1, HEK 293, Jurkat, and NIH 3T3 cells.

Quality and performance tested
Recovery Cell Culture Freezing Medium is quality tested for pH, osmolality, sterility, and endotoxin. In addition, each lot is performance tested using CHO-K1 cells.

cGMP manufacturing and quality system
For supply chain continuity, we manufacture Gibco Recovery Cell Culture Freezing Medium at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Culture TypeMammalian Cell Culture
FormLiquid
Product TypeFreezing Medium
Serum LevelStandard Serum Supplementation
Shelf Life12 Months
SterilitySterile-filtered
With AdditivesHigh Glucose, Phenol Red, DMSO (10%)
Without AdditivesNo Phenol Red
Product LineGibco™, Recovery™
Purity or Quality GradeResearch Grade
Quantity50 mL
Shipping ConditionDry Ice
Unit Size50 mL
Contents & Storage
Storage conditions: -5 to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture