2-Methyl-2-butene, 99+%
2-Methyl-2-butene, 99+%
2-Methyl-2-butene, 99+%
2-Methyl-2-butene, 99+%
Thermo Scientific Chemicals

2-Methyl-2-butene, 99+%

2-Methyl-2-butene, 99+%, C5H10, CAS Number-513-35-9, 1,1,2-trimethylethylene, 2-butene, 2-methyl, 2-methyl-2-butene, 2-methylbutene-2, 3-methyl-2-butene, amylene, beta-isoamylene, ethylene, trimethyl, n-amylene, trimethylethylene, 250mL, 99% min. (GC), 01, 211, 35.0 deg.C to 38.0 deg.C, 100.0, 99.0 | CAS: 513-35-9 | C5H10 | 70.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
Packaging:
Glass bottle
Catalog number 126492500
Price (EUR)
176,80
Online Exclusive
208,00
Save 31,20 (15%)
Each
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
176,80
Online Exclusive
208,00
Save 31,20 (15%)
Each
Add to cart
Chemical Identifiers
CAS513-35-9
IUPAC Name2-methylbut-2-ene
Molecular FormulaC5H10
InChI KeyBKOOMYPCSUNDGP-UHFFFAOYSA-N
SMILESCC=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3860 to 1.3880 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link