4-Methylcatechol, 98%, Thermo Scientific Chemicals
4-Methylcatechol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylcatechol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 126511000
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.10 
Save 10.45 (14%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.10 
Save 10.45 (14%)
Add to cart
4-Methylcatechol, 98%, Thermo Scientific Chemicals
Catalog number126511000
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.10 
Save 10.45 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-methylbenzene-1,2-diol
Molecular FormulaC7H8O2
InChI KeyZBCATMYQYDCTIZ-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(O)=C1
Molecular Weight (g/mol)124.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Melting point63°C to 68°C
Appearance (Color)White to brown
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only