Thermo Scientific Chemicals

Methyl chloroformate, 99%, Thermo Scientific Chemicals

Catalog number: 126530050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methyl chloroformate, 99%, Thermo Scientific Chemicals

Catalog number: 126530050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 126530050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-22-1
IUPAC Name
methyl carbonochloridate
Molecular Formula
C2H3ClO2
InChI Key
XMJHPCRAQCTCFT-UHFFFAOYSA-N
SMILES
COC(Cl)=O
Appearance (Color)
Clear colorless to light yellow
GC
>=98.5 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.3860 to 1.3880 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text