Thermo Scientific Chemicals

Methylcyclohexane, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 126580025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

Methylcyclohexane, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 126580025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 126580025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-87-2
IUPAC Name
methylcyclohexane
Molecular Formula
C7H14
InChI Key
UAEPNZWRGJTJPN-UHFFFAOYSA-N
SMILES
CC1CCCCC1
Color scale
=<20 APHA
Refractive index
1.4210 to 1.4240 (20°C, 589 nm)
Residue after evaporation
=<0.01 %
Infrared spectrum
Conforms
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text