Methylcyclohexane, 99%, extra pure, Thermo Scientific Chemicals
Methylcyclohexane, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylcyclohexane, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 126580050
Price (USD)/ Each
261.65
Online exclusive
291.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
261.65
Online exclusive
291.00 
Save 29.35 (10%)
Add to cart
Methylcyclohexane, 99%, extra pure, Thermo Scientific Chemicals
Catalog number126580050
Price (USD)/ Each
261.65
Online exclusive
291.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS108-87-2
IUPAC Namemethylcyclohexane
Molecular FormulaC7H14
InChI KeyUAEPNZWRGJTJPN-UHFFFAOYSA-N
SMILESCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Refractive index1.4210 to 1.4240 (20°C, 589 nm)
Residue after evaporation=<0.01 %
Infrared spectrumConforms
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only