Informational:Detect rhabdovirus contamination in Sf9 cultures. Learn more »
Informational:Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™

Sf-900™ III SFM

Catalog number: 12658019
Have Questions?
Gibco™

Sf-900™ III SFM

Catalog number: 12658019
Have Questions?
Catalog Number
12658019
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
12658019500 mL
-
126580271,000 mL
-
1265803510 L
-
1265800120 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Sf-900™ III SFM is a low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells and for recombinant gene expression using the baculovirus and stable insect expression systems. This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines. Features of Gibco™ Sf-900™ III SFM:

• Superior long-term, high-density growth
• Optimized for recombinant protein production
• Serum-free, protein-free, animal origin–free, ready-to-use formulation
• Improved lot-to-lot consistency

Superior long-term, high-density growth
Spodoptera frugiperda (Sf9) cells grown in Sf-900™ III SFM achieve maximum cell densities of 10 to 14 × 106 cells/mL, a significant improvement over Sf-900™ II SFM and other commercially available serum-free media (see figure).

Optimized for recombinant protein production
Traditionally, Grace's medium supplemented with 10% FBS has been used for recombinant protein expression. Sf-900™ III SFM is an improved serum-free, protein-free, animal origin–free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect viruses and recombinant proteins.

Serum-free, protein-free, animal origin–free, ready-to-use formulation
Gibco™ Sf-900™ III SFM is a serum-free, protein-free, animal origin–free medium that allows for much easier purification of your protein of interest. Sf-900™ III SFM is ready to use; it does not require addition of serum, glutamine, or surfactants. Cells adapted to other commercially available serum-free media should be sequentially adapted into Sf-900™ III SFM (see product manual for details).

Improved lot-to-lot consistency
Gibco™ Sf-900™ III SFM contains reduced hydrolysate concentrations, improving lot-to-lot consistency over Sf-900™ II SFM (see figure).

cGMP manufacturing and quality system
Gibco™ Sf-900 II SFM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line
Sf21, Sf9
Cell Type
Insect Cell
Classification
Animal Origin-free, Protein-free, Serum-free
Form
Liquid
Product Type
Insect Cell Serum Free Medium (SFM)
Serum Level
Serum-free
With Additives
Glutamine
Green Features
Sustainable packaging
Product Line
Gibco™, Sf-900™
Quantity
500 mL
Shipping Condition
Room Temperature
Species
S. frugiperda, Spodoptera frugiperda

Contents & Storage

Storage conditions: 2–8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text