Protein Expression Medium (PEM)
Protein Expression Medium (PEM)
Gibco™

Protein Expression Medium (PEM)

• Supports the large-scale, high-density culture of transfected PER.C6™ cells in shake flasks, roller bottles, and bioreactors• Contains no componentsRead more
Have Questions?
Catalog NumberQuantity
126610131 L
Catalog number 12661013
Price (USD)
146.00
Each
-
Add to cart
Quantity:
1 L
Price (USD)
146.00
Each
Add to cart
• Supports the large-scale, high-density culture of transfected PER.C6™ cells in shake flasks, roller bottles, and bioreactors
• Contains no components directly derived from human or animal sources and one low-level, recombinant protein, minimizing the opportunity to introduce adventitious agents
• Formulated without L-glutamine for maximum stability
• Supports parental PER.C6™, HEK 293, and HeLa cell growth in suspension culture, as well as transfected PER.C6™ cells for protein expression applications Storage: Store in the dark at 2-8°C. PER.C6™ is a registered trademark of Crucell, Holland B.V.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineHeLa, PER.C6, 293 (HEK)
Concentration1 X
Endotoxin LevelLow
FormatBottle(s)
Included AntibioticsNo antibiotics
Product LineGibco™
Product TypeProtein Expression Medium (PEM)
Quantity1 L
Shelf Life12 Months
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Low Protein, Serum-free
Culture TypeSuspension Cell Culture
FormLiquid
Serum LevelSerum-free
With AdditivesSodium Bicarbonate
Without AdditivesNo Glutamine, No Phenol Red, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link