Thermo Scientific Chemicals

4,4'-Methylenebis(N,N-dimethylaniline), 98%, Thermo Scientific Chemicals

Catalog number: 126741000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4,4'-Methylenebis(N,N-dimethylaniline), 98%, Thermo Scientific Chemicals

Catalog number: 126741000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 126741000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101-61-1
IUPAC Name
4-{[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
Molecular Formula
C17H22N2
InChI Key
JNRLEMMIVRBKJE-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(CC2=CC=C(C=C2)N(C)C)C=C1
Appearance (Color)
White to beige to grey to brown
Infrared spectrum
Conforms
Melting point
86°C to 92°C
Appearance (Form)
Powder or crystals and/or chunks
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text