LHC-8 Medium (1X), without gentamicin
Product Image
Gibco™

LHC-8 Medium (1X), without gentamicin

LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. This is a supplemented medium.Read more
Have Questions?
Catalog NumberQuantity
12679015500 mL
Catalog number 12679015
Price (EUR)
173,00
Each
-
Add to cart
Quantity:
500 mL
Price (EUR)
173,00
Each
Add to cart
Ask our AI about this Product
LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. This is a supplemented medium. For maximum flexibility and convenience, we also offer LHC basal medium.

Applications:
Engineered specifically for the growth of bronchial epithelial cells such as IB3-1, S9, and C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), and BBE (bovine bronchial epithelial cells) for asthma, allergy, lung cancer, cystic fibrosis, pulmonary, and esophageal cancer research without further supplements.

Performance and quality testing:
Performance tested in a growth assay using BEAS-2B cells.

Reference(s):
Pawliczak R., et. al. (2001) J Biol Chem 276, 44613. Zhao Y.L., et. al. (2000) Carcinogenesis 21, 2005. Liu X., et. al. (1998) Am J Physiol 274, L58. Hei T.K., et. al. (1997) Environ Health Perspect 105, 1085. Lechner J.F., and LaVeck, M.A. (1985) J Tissue Cult Methods 9, 43.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineB3-1, S9, C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), BBE (bovine bronchial epithelial cells)
Cell TypeEpithelial Cells
Concentration1 X
Included AntibioticsNo Gentamicin Sulfate
Product TypeLHC-8 Medium
Quantity500 mL
Shelf Life12 Months
Shipping ConditionDry Ice
ClassificationSerum-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Without AdditivesNo Gentamicin
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection