Thermo Scientific Chemicals

N-Methylformanilide, 99%, Thermo Scientific Chemicals

Catalog number: 126810100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

N-Methylformanilide, 99%, Thermo Scientific Chemicals

Catalog number: 126810100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 126810100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
93-61-8
IUPAC Name
N-methyl-N-phenylformamide
Molecular Formula
C8H9NO
InChI Key
JIKUXBYRTXDNIY-UHFFFAOYSA-N
SMILES
CN(C=O)C1=CC=CC=C1
GC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Refractive index
1.5600 to 1.5620 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text