N-Methylformanilide, 99%, Thermo Scientific Chemicals
N-Methylformanilide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methylformanilide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 126815000
Price (USD)/ Each
81.90
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
81.90
Add to cart
N-Methylformanilide, 99%, Thermo Scientific Chemicals
Catalog number126815000
Price (USD)/ Each
81.90
-
Add to cart
Chemical Identifiers
CAS93-61-8
IUPAC NameN-methyl-N-phenylformamide
Molecular FormulaC8H9NO
InChI KeyJIKUXBYRTXDNIY-UHFFFAOYSA-N
SMILESCN(C=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Refractive index1.5600 to 1.5620 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only