Thermo Scientific Chemicals

Methyl formate, 97%, pure, Thermo Scientific Chemicals

Catalog number: 126820100
10 L, Metal drum, Each
Thermo Scientific Chemicals

Methyl formate, 97%, pure, Thermo Scientific Chemicals

Catalog number: 126820100
10 L, Metal drum, Each
Quantity
Packaging
Catalog number: 126820100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-31-3
IUPAC Name
methyl formate
Molecular Formula
C2H4O2
InChI Key
TZIHFWKZFHZASV-UHFFFAOYSA-N
SMILES
COC=O
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Water
=<0.05 %
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text