3-Methylglutaric anhydride, 97%, Thermo Scientific Chemicals
3-Methylglutaric anhydride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylglutaric anhydride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 126870250
Price (USD)/ Each
166.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
166.00
Add to cart
3-Methylglutaric anhydride, 97%, Thermo Scientific Chemicals
Catalog number126870250
Price (USD)/ Each
166.00
-
Add to cart
Chemical Identifiers
CAS4166-53-4
IUPAC Name4-methyloxane-2,6-dione
Molecular FormulaC6H8O3
InChI KeyMGICRVTUCPFQQZ-UHFFFAOYSA-N
SMILESCC1CC(=O)OC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Assay>=96.0 %
Appearance (Form)Low melting crystalline solid
Melting point40°C to 45°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only