Methyl 4-hydroxybenzoate, 99%
Methyl 4-hydroxybenzoate, 99%
Methyl 4-hydroxybenzoate, 99%
Methyl 4-hydroxybenzoate, 99%
Thermo Scientific Chemicals

Methyl 4-hydroxybenzoate, 99%

CAS: 99-76-3 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
10 kg
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 126960025
Price (USD)
359.00
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
359.00
Each
Add to cart
Chemical Identifiers
CAS99-76-3
IUPAC Namemethyl 4-hydroxybenzoate
Molecular FormulaC8H8O3
InChI KeyLXCFILQKKLGQFO-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point125°C to 128°C
Appearance (Color)White to almost white
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link