Methyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Methyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
100 g
500 g
2.5 kg
Packaging:
View all
Plastic Drum
Catalog number 126960100
Price (USD)/ Each
850.65
Online exclusive
1,001.00 
Save 150.35 (15%)
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
850.65
Online exclusive
1,001.00 
Save 150.35 (15%)
Add to cart
Methyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Catalog number126960100
Price (USD)/ Each
850.65
Online exclusive
1,001.00 
Save 150.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namemethyl 4-hydroxybenzoate
Molecular FormulaC8H8O3
InChI KeyLXCFILQKKLGQFO-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(O)C=C1
Molecular Weight (g/mol)152.15
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point125°C to 128°C
Appearance (Color)White to almost white
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only