Thermo Scientific Chemicals

Methyl 2-hydroxyisobutyrate, 99%, Thermo Scientific Chemicals

Catalog number: 126970500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 2-hydroxyisobutyrate, 99%, Thermo Scientific Chemicals

Catalog number: 126970500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 126970500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2110-78-3
IUPAC Name
methyl 2-hydroxy-2-methylpropanoate
Molecular Formula
C5H10O3
InChI Key
XYVQFUJDGOBPQI-UHFFFAOYSA-N
SMILES
COC(=O)C(C)(C)O
Appearance (Form)
Liquid
GC
>=98.5 %
Infrared spectrum
Conforms
Refractive index
1.4100 to 1.4120 (20°C, 589 nm)
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text