2-Methylimidazole, 99%, Thermo Scientific Chemicals
2-Methylimidazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylimidazole, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 127001000
Price (USD)/ Each
29.65
Online exclusive
32.60 
Save 2.95 (9%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
29.65
Online exclusive
32.60 
Save 2.95 (9%)
Add to cart
2-Methylimidazole, 99%, Thermo Scientific Chemicals
Catalog number127001000
Price (USD)/ Each
29.65
Online exclusive
32.60 
Save 2.95 (9%)
-
Add to cart
Chemical Identifiers
CAS693-98-1
IUPAC Name2-methyl-1H-imidazole
Molecular FormulaC4H6N2
InChI KeyLXBGSDVWAMZHDD-UHFFFAOYSA-N
SMILESCC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Water=<0.5 % (K.F.)
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only