N-Methyliminodiacetic acid, 99%, Thermo Scientific Chemicals
N-Methyliminodiacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methyliminodiacetic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 127010250
Price (USD)/ Each
179.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
179.00
Add to cart
N-Methyliminodiacetic acid, 99%, Thermo Scientific Chemicals
Catalog number127010250
Price (USD)/ Each
179.00
-
Add to cart
Chemical Identifiers
CAS4408-64-4
IUPAC Name2-[(carboxymethyl)(methyl)amino]acetic acid
Molecular FormulaC5H9NO4
InChI KeyXWSGEVNYFYKXCP-UHFFFAOYSA-N
SMILESCN(CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only