Thermo Scientific Chemicals

2-Methylindole, 98%, Thermo Scientific Chemicals

Catalog number: 127021000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Methylindole, 98%, Thermo Scientific Chemicals

Catalog number: 127021000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 127021000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
95-20-5
IUPAC Name
2-methyl-1H-indole
Molecular Formula
C9H9N
InChI Key
BHNHHSOHWZKFOX-UHFFFAOYSA-N
SMILES
CC1=CC2=CC=CC=C2N1
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
56°C to 60°C
Appearance (Form)
Crystalline powder or crystals or flakes
Appearance (Color)
Yellow to red-purple to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text