3-Methylindole, 98%, Thermo Scientific Chemicals
3-Methylindole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylindole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 127030100
Price (USD)/ Each
45.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
53.50 
Save 7.85 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
53.50 
Save 7.85 (15%)
Add to cart
3-Methylindole, 98%, Thermo Scientific Chemicals
Catalog number127030100
Price (USD)/ Each
45.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
53.50 
Save 7.85 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name3-methyl-1H-indole
Molecular FormulaC9H9N
InChI KeyZFRKQXVRDFCRJG-UHFFFAOYSA-N
SMILESCC1=CNC2=CC=CC=C12
Molecular Weight (g/mol)131.18
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or flakes
GC>=97.5 %
Appearance (Color)White to light brown
Melting point95°C to 98°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only