3-Methylindole, 98%, Thermo Scientific Chemicals
3-Methylindole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylindole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 127030250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS83-34-1
IUPAC Name3-methyl-1H-indole
Molecular FormulaC9H9N
InChI KeyZFRKQXVRDFCRJG-UHFFFAOYSA-N
SMILESCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or flakes
GC>=97.5 %
Appearance (Color)White to light brown
Melting point95°C to 98°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only