Thermo Scientific Chemicals

3-Methylindole, 98%, Thermo Scientific Chemicals

Catalog number: 127031000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

3-Methylindole, 98%, Thermo Scientific Chemicals

Catalog number: 127031000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 127031000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
83-34-1
IUPAC Name
3-methyl-1H-indole
Molecular Formula
C9H9N
InChI Key
ZFRKQXVRDFCRJG-UHFFFAOYSA-N
SMILES
CC1=CNC2=CC=CC=C12
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or flakes
GC
>=97.5 %
Appearance (Color)
White to light brown
Melting point
95°C to 98°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text