Methyl isonicotinate, 98%, Thermo Scientific Chemicals
Methyl isonicotinate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl isonicotinate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number 127065000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2459-09-8
IUPAC Namemethyl pyridine-4-carboxylate
Molecular FormulaC7H7NO2
InChI KeyOLXYLDUSSBULGU-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear orange to brown
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5110 to 1.5140 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only