Methylmalonic acid, 96%, Thermo Scientific Chemicals
Methylmalonic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylmalonic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 127091000
Price (USD)/ Each
589.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
589.00
Add to cart
Methylmalonic acid, 96%, Thermo Scientific Chemicals
Catalog number127091000
Price (USD)/ Each
589.00
-
Add to cart
Chemical Identifiers
IUPAC Name2-methylpropanedioic acid
Molecular FormulaC4H6O4
InChI KeyZIYVHBGGAOATLY-UHFFFAOYSA-N
SMILESCC(C(O)=O)C(O)=O
Molecular Weight (g/mol)118.09
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=95 %
Appearance (Color)White
Water=<0.5 % (K.F.)
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only