2-Methylpentane, 99+%, pure
2-Methylpentane, 99+%, pure
2-Methylpentane, 99+%, pure
2-Methylpentane, 99+%, pure
Thermo Scientific Chemicals

2-Methylpentane, 99+%, pure

CAS: 107-83-5 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 127341001
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-83-5
IUPAC Name2-methylpentane
Molecular FormulaC6H14
InChI KeyAFABGHUZZDYHJO-UHFFFAOYSA-N
SMILESCCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
GC>=99.0 %
Refractive index1.3700 to 1.3720 (20°C, 589 nm)
Appearance (Form)Liquid
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467762Certificate of AnalysisJan 30, 2025127341001, 127340010, 127340025
A0459384Certificate of AnalysisJan 17, 2024127341001, 127340010, 127340025
A0430332Certificate of AnalysisNov 20, 2023127341001, 127340010, 127340025
A0430331Certificate of AnalysisNov 17, 2023127341001, 127340010, 127340025
A0402676Certificate of AnalysisDec 15, 2022127341001, 127340010, 127340025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection