3-Methylpentane, 99+%, Thermo Scientific Chemicals
3-Methylpentane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylpentane, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 127350500
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.10 
Save 6.45 (14%)
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.10 
Save 6.45 (14%)
Add to cart
3-Methylpentane, 99+%, Thermo Scientific Chemicals
Catalog number127350500
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.10 
Save 6.45 (14%)
-
Add to cart
Chemical Identifiers
CAS96-14-0
IUPAC Name3-methylpentane
Molecular FormulaC6H14
InChI KeyPFEOZHBOMNWTJB-UHFFFAOYSA-N
SMILESCCC(C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3755 to 1.3775 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only