3-Methylpentane, 99+%
3-Methylpentane, 99+%
3-Methylpentane, 99+%
3-Methylpentane, 99+%
Thermo Scientific Chemicals

3-Methylpentane, 99+%

CAS: 96-14-0 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
1 L
250 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 127352500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS96-14-0
IUPAC Name3-methylpentane
Molecular FormulaC6H14
InChI KeyPFEOZHBOMNWTJB-UHFFFAOYSA-N
SMILESCCC(C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3755 to 1.3775 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection