1x1 image pixel for data collection
3-Methyl-2-pentanone, 99%
3-Methyl-2-pentanone, 99%
3-Methyl-2-pentanone, 99%
3-Methyl-2-pentanone, 99%
Thermo Scientific Chemicals

3-Methyl-2-pentanone, 99%

CAS: 565-61-7 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 127380250
Price (USD)
211.65
Online Exclusive
235.00
Save 23.35 (10%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
211.65
Online Exclusive
235.00
Save 23.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS565-61-7
IUPAC Name3-methylpentan-2-one
Molecular FormulaC6H12O
InChI KeyUIHCLUNTQKBZGK-UHFFFAOYNA-N
SMILESCCC(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
GC>=98.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection