N-Methylphenethylamine, 99%, Thermo Scientific Chemicals
N-Methylphenethylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methylphenethylamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 127450050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namemethyl(2-phenylethyl)amine
Molecular FormulaC9H13N
InChI KeySASNBVQSOZSTPD-UHFFFAOYSA-N
SMILESCNCCC1=CC=CC=C1
Molecular Weight (g/mol)135.21
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5150 to 1.5170 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only