3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%, Thermo Scientific Chemicals
3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 127460050
Price (USD)/ Each
30.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
30.00
Add to cart
3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%, Thermo Scientific Chemicals
Catalog number127460050
Price (USD)/ Each
30.00
-
Add to cart
Chemical Identifiers
IUPAC Name3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Molecular FormulaC10H10N2O
InChI KeyQELUYTUMUWHWMC-UHFFFAOYSA-N
SMILESCC1=NN(C(=O)C1)C1=CC=CC=C1
Molecular Weight (g/mol)174.20
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point127°C to 131°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Appearance (Color)Yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only