Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%, Thermo Scientific Chemicals

Catalog number: 127465000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%, Thermo Scientific Chemicals

Catalog number: 127465000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 127465000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-25-8
IUPAC Name
3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Molecular Formula
C10H10N2O
InChI Key
QELUYTUMUWHWMC-UHFFFAOYSA-N
SMILES
CC1=NN(C(=O)C1)C1=CC=CC=C1
Melting point
127°C to 131°C
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
HPLC
>=98.5 %
Appearance (Color)
Yellow to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text