N-Methyl-N-propargylbenzylamine, 97%, Thermo Scientific Chemicals
N-Methyl-N-propargylbenzylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methyl-N-propargylbenzylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 127530010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namebenzyl(methyl)(prop-2-yn-1-yl)amine
Molecular FormulaC11H13N
InChI KeyDPWPWRLQFGFJFI-UHFFFAOYSA-N
SMILESCN(CC#C)CC1=CC=CC=C1
Molecular Weight (g/mol)159.23
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=96.0 %
Refractive index1.5210 to 1.5230 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only