Thermo Scientific Chemicals

N-Methylpyrrole, 99+%, Thermo Scientific Chemicals

Catalog number: 127605000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Methylpyrrole, 99+%, Thermo Scientific Chemicals

Catalog number: 127605000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 127605000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
96-54-8
IUPAC Name
1-methyl-1H-pyrrole
Molecular Formula
C5H7N
InChI Key
OXHNLMTVIGZXSG-UHFFFAOYSA-N
SMILES
CN1C=CC=C1
GC
>=99.0 %
Refractive index
1.4860 to 1.4890 (20°C, 589 nm)
Appearance (Form)
Liquid
Water
=<0.5 % (K.F.)
Appearance (Color)
Clear yellow to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text