Thermo Scientific Chemicals

1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals

Catalog number: 127615000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals

Catalog number: 127615000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 127615000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
120-94-5
IUPAC Name
1-methylpyrrolidine
Molecular Formula
C5H11N
InChI Key
AVFZOVWCLRSYKC-UHFFFAOYSA-N
SMILES
CN1CCCC1
GC
>=97.5 %
Appearance (Form)
Liquid
Refractive index
1.4230 to 1.4260 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text