1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99%, extra pure

1-Methyl-2-pyrrolidinone, 99%, C5H9NO, CAS Number-872-50-4, 1-methyl-2-pyrrolidinone, 1-methyl-2-pyrrolidone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidinone, n-methylpyrrolidone, 2.5L | CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
25 L
1 L
Packaging:
View all
Glass Bottle
Catalog number 127630010
Price (USD)
92.65
Special offer
Online exclusive
Ends: 26-Sep-2025
109.00
Save 16.35 (15%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 26-Sep-2025
109.00
Save 16.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Water=<0.05 % (K.F.)
Infrared spectrumConforms
Specific gravity(20°C) 1.02 to 1.04
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link