1x1 image pixel for data collection
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
1-Methyl-2-pyrrolidinone, 99%, extra pure
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99%, extra pure

1-Methyl-2-pyrrolidinone, 99%, C5H9NO, CAS Number-872-50-4, 1-methyl-2-pyrrolidinone, 1-methyl-2-pyrrolidone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidinone, n-methylpyrrolidone, 2.5L | CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
25 L
1 L
Packaging:
View all
Glass Bottle
Catalog number 127630025
Price (USD)
188.65
Special offer
Online exclusive
Ends: 26-Sep-2025
222.00
Save 33.35 (15%)
Each
In stock
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
188.65
Special offer
Online exclusive
Ends: 26-Sep-2025
222.00
Save 33.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Water=<0.05 % (K.F.)
Infrared spectrumConforms
Specific gravity(20°C) 1.02 to 1.04
View more
Ask our AI about this Product

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0474356Certificate of AnalysisJul 10, 2025127630025, 127630250, 127630010
A0470403Certificate of AnalysisMay 17, 2025127630025, 127630250, 127630010
A0435139Certificate of AnalysisOct 07, 2024127630025, 127630250, 127630010, 12763025
A0463748Certificate of AnalysisJun 24, 2024127630025, 127630250, 127630010, 12763025
A0460628Certificate of AnalysisFeb 27, 2024127630025, 127630250, 127630010, 12763025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection