Methyl pyruvate, 94%, Thermo Scientific Chemicals
Methyl pyruvate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl pyruvate, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 127655000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namemethyl 2-oxopropanoate
Molecular FormulaC4H6O3
InChI KeyCWKLZLBVOJRSOM-UHFFFAOYSA-N
SMILESCOC(=O)C(C)=O
Molecular Weight (g/mol)102.09
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=92 %
Refractive index1.4035 to 1.4075 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only