alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 127710025
Price (USD)/ Each
98.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
116.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
98.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
116.00 
Save 17.35 (15%)
Add to cart
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
Catalog number127710025
Price (USD)/ Each
98.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
116.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(prop-1-en-2-yl)benzene
Molecular FormulaC9H10
InChI KeyXYLMUPLGERFSHI-UHFFFAOYSA-N
SMILESCC(=C)C1=CC=CC=C1
Molecular Weight (g/mol)118.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to almost colorless
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5360 to 1.5400 (20°C, 589 nm)
Stabilizer10 to 20 ppm tert-Butylcatechol
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only