Thermo Scientific Chemicals

Methyl sulfone, 98%, Thermo Scientific Chemicals

Catalog number: 127785000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Methyl sulfone, 98%, Thermo Scientific Chemicals

Catalog number: 127785000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 127785000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
67-71-0
IUPAC Name
methanesulfonylmethane
Molecular Formula
C2H6O2S
InChI Key
HHVIBTZHLRERCL-UHFFFAOYSA-N
SMILES
CS(C)(=O)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Melting point
107°C to 111°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text