2-Methyl-2-thiopseudourea hemisulfate, 98%, Thermo Scientific Chemicals
2-Methyl-2-thiopseudourea hemisulfate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-2-thiopseudourea hemisulfate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 127891000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS867-44-7
IUPAC Namebis((methylsulfanyl)methanimidamide); sulfuric acid
Molecular FormulaC4H14N4O4S3
InChI KeyBZZXQZOBAUXLHZ-UHFFFAOYSA-N
SMILESCSC(N)=N.CSC(N)=N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only