Thermo Scientific Chemicals

Methyltrichlorosilane, 98+%, Thermo Scientific Chemicals

Catalog number: 127930100
10 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methyltrichlorosilane, 98+%, Thermo Scientific Chemicals

Catalog number: 127930100
10 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 127930100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-79-6
IUPAC Name
trichloro(methyl)silane
Molecular Formula
CH3Cl3Si
InChI Key
JLUFWMXJHAVVNN-UHFFFAOYSA-N
SMILES
C[Si](Cl)(Cl)Cl
GC
>=98.0 %
Appearance (Form)
Liquid
Refractive index
1.4100 to 1.4120 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text