Methyltrichlorosilane, 98+%
Methyltrichlorosilane, 98+%
Methyltrichlorosilane, 98+%
Methyltrichlorosilane, 98+%
Thermo Scientific Chemicals

Methyltrichlorosilane, 98+%

CAS: 75-79-6 | CH3Cl3Si | 149.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
500 mL
Packaging:
Glass bottle
Catalog number 127935000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-79-6
IUPAC Nametrichloro(methyl)silane
Molecular FormulaCH3Cl3Si
InChI KeyJLUFWMXJHAVVNN-UHFFFAOYSA-N
SMILESC[Si](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.0 %
Appearance (Form)Liquid
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link