Thermo Scientific Chemicals

5-Methyl-DL-tryptophan, 98%

Catalog number: 127970010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Methyl-DL-tryptophan, 98%

Catalog number: 127970010
1 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 127970010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
951-55-3
IUPAC Name
2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
Molecular Formula
C12H14N2O2
InChI Key
HUNCSWANZMJLPM-UHFFFAOYSA-N
SMILES
CC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1
Appearance (Form)
Powder
Heavy metals (as Pb)
=<10 ppm
Appearance of solution
(5% soln. in 1 N NaOH) clear, pale yellowish
Infrared spectrum
Conforms
Water
=<1 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text