DMEM, powder, high glucose, pyruvate
Save up to 31% on your order of cell culture products. View promotion details »
DMEM, powder, high glucose, pyruvate
Gibco™

DMEM, powder, high glucose, pyruvate

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1280008210 L
1280001710 x 1 L
Catalog number 12800082
Price (USD)/ 10 L
42.08
-
Add to cart
Quantity:
10 L
Customize this product
Price (USD)/ 10 L
42.08
Add to cart
DMEM, powder, high glucose, pyruvate
Catalog number12800082
Price (USD)/ 10 L
42.08
-
Add to cart

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This DMEM is modified as follows:
WithWithout
• High Glucose• HEPES
• L-glutamine• Sodium Bicarbonate
• Phenol Red 
• Sodium Pyruvate 


The complete formulation is available.

Using DMEM
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture media require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

In vitro diagnostic
Specifications
Cell LineHeLa, 293, Cos-7, and PC-12
Cell TypePrimary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
ClassificationAnimal Origin-free
FormPowder
Product TypeDMEM (Dulbecco's Modified Eagle Medium)
Serum LevelStandard Serum Supplementation
Shelf Life36 Months From Date of Manufacture
SterilitySterile-filtered
With AdditivesHigh Glucose, Glutamine, Phenol Red, Sodium Pyruvate
Without AdditivesNo HEPES, No Sodium Bicarbonate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco™
Quantity10 L
Shipping ConditionRoom Temperature
Unit Size10 L
Contents & Storage
Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture